This product is now discontinued, View Related Products
Argos Technologies® Polypropylene Freezer Storage Boxes
Featuring reinforced lock and seal for secure storage
  • Reinforced triple hinges prevent cracks or breaks
  • Bold, black alpha-numeric printing allows accurate tracking of samples
  • Click fit lock ensures box will not open accidentally -- large clasp allows easy opening with gloves
Boxes withstand temperatures from -80°C to 121°C, and will not degrade after repeated thaws. Each box features an alphanumeric grid on the exterior and interior, stacking rings and a matte surface suitable for markers or labels. Available with clear or fluorescent colored lids with clear bottoms for easy visual identification. Boxes accommodate 0.5, 1.5, and 2.0 mL microtubes and most brands of cryogenic vials. Black box is suitable for light sensitive samples.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Color Description
Availability Pricing
COMPARE
Black Black Freezer Box, 100 Place, Polypropylene, 50/cs
Call For Price
COMPARE
Clear Clear Freezer Boxes, 100 Place, Polypropylene, 50/cs
Call For Price
COMPARE
Blue Blue Freezer Boxes, 100 Place, Polypropylene, 50/cs
Call For Price
COMPARE
Green Green Freezer Boxes, 100 Place, Polypropylene, 50/cs
Call For Price
COMPARE
Orange Orange Freezer Boxes, 100 Place, Polypropylene, 50/cs
Call For Price
COMPARE
Purple Purple Freezer Boxes, 100 Place, Polypropylene, 50/cs
Call For Price
COMPARE
Assorted Assorted Color Freezer Boxes, 100 Place, PP, 50/cs
Call For Price
Showing 1- 7 of 7