This product is now discontinued, View Related Products
Argos Technologies® PolarSafe® Vertical/Chest Aluminum Racks for Mini 3" Boxes
Aluminum racks offer better temperature uniformity in vapor phase in liquid nitrogen tanks
  • Lightweight aluminum construction for easier handling
  • Use with a wide variety of chest freezers and liquid nitrogen tanks
  • Accommodates mini (3" x 3" x 3") cardboard or plastic boxes
  • Locking rod keeps sample storage boxes securely in the rack
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
7 boxes 22 1/4
Call For Price
COMPARE
8 boxes 25 7/16
Call For Price
COMPARE
9 boxes 28 9/16
Call For Price
COMPARE
10 boxes 31 3/4