This product is now discontinued, View Related Products
Argos Technologies® HDPE Cryogenic Storage Dividers
Great for use in standard 3" boxes or directly on the benchtop!
  • Rigid High Density Polyethylene (HDPE) construction
  • Choose from 25-place or 81-place dividers
Argos Technologies tube divider is great for organizing and storing microcentrifuge tubes either on your benchtop or in the Argos Technologies standard 3” cardboard freezer box (R3016A). Designed to accommodate the various tube designs on the market and each of the wells are numbered for quick sample identification. Constructed of rigid HDPE, the divider features a textured post on top for easy handling. Unit measures 4.9” x 4.9” x 2.75” (12.5 x 12.5 x 6.9 cm) and handles temperatures to 248°F (120°C).
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Tubes Held Tube Size Description
Availability Pricing
COMPARE
25 5 mL Plastic Freezer Box Divider, 25-Place, for 5.0 mL Tubes
Call For Price
COMPARE
81 1.5 to 2.0 mL Plastic Freezer Box Divider, 81-Place, for 1.5 to 2.0 mL Tubes; 5/Pk