Cole-Parmer® Essentials Cell Scrapers with Pivoting Blade, Sterile
Cell scraping tool makes harvesting cells from flasks, dishes, and plates easy
  • Pivoting blade helps to minimize cell damage
  • Ribbed-grip design prevents slippage during use
  • Sterile, individually packaged
  • Nonpyrogenic
Designed to make the process of scraping and collecting cells easier and more effective, these irradiation sterilized (SAL 10-6) cell scrapers have an ultra-thin, pivoting TPE blade on an ABS plastic handle. The blade ensures an ideal angle to allow access to hard-to-reach areas and helps reduce cell damage. The handle has ribs to ensure an easy, no-slip grip.
Manufactured under an ISO 13485 and ISO 9001 quality management system in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these cell scrapers are DNase/RNase free, nonpyrogenic, noncytotoxic, and conform to USP CLASS VI.
£141.17 - £153.91GBP / Case of 100
£169.40 - £184.69GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Overall Length (mm) Sterile Description
Availability Pricing
COMPARE
250 Yes Cell Scraper, Sterile, 20 mm Pivoting Blade, 25 cm L; 100/Cs
£141.17GBP / Case of 100
£169.40GBP (incl VAT)
COMPARE
390 Yes Cell Scraper, Sterile, 30 mm Pivoting Blade, 39 cm L; 100/Cs
£153.91GBP / Case of 100
£184.69GBP (incl VAT)