Cole-Parmer® Essentials Sterile Lab Sampling Scoops, Polystyrene
Sterile, disposable lab scoops prevent cross contamination
  • Meets FDA guidelines for food, drug and cosmetic sampling
  • Flat bottom for use as weigh boat
  • Ergonomic handle for a comfortable grip
  • Individually sealed in easy-to-open bags
Use these sterile, disposable lab scoops to help reduce cross contamination. Manufactured from sturdy, one-piece, FDA-grade polystyrene, they can be used to hold a variety of media and have an ergonomic, comfortably curved handle to make sample retrieval easy. A flat bottom allows them to be placed on a surface without spillage and to be used as a weighing dish.
These individually bagged laboratory scoops are gamma irradiation sterilized SAL 10-6; free of BSE/TSE, heavy metals, BPA, and phthalates; and lot traced. A Certificate of Irradiation is available upon request. Specified volumes are approximate and not intended for measuring specific volumes.
£114.98 - £193.30GBP / Case of 100
£137.98 - £231.96GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (cm) Width (cm)
Availability Pricing
COMPARE
30 18 3.4
£114.98GBP / Case of 100
£137.98GBP (incl VAT)
COMPARE
60 20.32 3.81
£126.40GBP / Case of 100
£151.68GBP (incl VAT)
COMPARE
120 22.86 5.08
£174.86GBP / Case of 100
£209.83GBP (incl VAT)
COMPARE
250 24.66 7.83
£193.30GBP / Case of 100
£231.96GBP (incl VAT)
COMPARE
500 30.44 10
£128.87GBP / Case of 50
£154.64GBP (incl VAT)
COMPARE
1000 36.26 12.15
£128.87GBP / Case of 25
£154.64GBP (incl VAT)