Cole-Parmer® Essentials Anti-Static Weighing Funnels, Polypropylene
Ensure complete sample transfer
  • Autoclavable
  • 6 month warranty
These disposable polypropylene (PP) weighing funnels are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance.
£87.00 - £117.80GBP / Pkg of 96
£104.40 - £141.36GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Stem OD (mm) Description
Availability Pricing
COMPARE
Polypropylene (PP) 6.21 Anti-Static Weighing Funnel, PP, Micro; 96/PK
£117.80GBP / Pkg of 96
£141.36GBP (incl VAT)
COMPARE
Polypropylene (PP) 7.4 Anti-Static Weighing Funnel, PP, Small; 50/PK
£93.07GBP / Pkg of 50
£111.68GBP (incl VAT)
COMPARE
Polypropylene (PP) 10.35 Anti-Static Weighing Funnel, PP, Medium; 50/PK
£105.00GBP / Pkg of 50
£126.00GBP (incl VAT)
COMPARE
Polypropylene (PP) 17 Anti-Static Weighing Funnel, PP, Large; 20/PK
£87.00GBP / Pkg of 20
£104.40GBP (incl VAT)
COMPARE
Polypropylene (PP) 19.1 Anti-Static Weighing Funnel, PP, Extra Large; 10/PK
£96.76GBP / Pkg of 10
£116.11GBP (incl VAT)