This product is now discontinued, View Related Products
Amprobe UAT-Series Underground Utility Locators
Accurately pinpoint underground utilities and buried services up with accurate depth measurement to 20 feet
  • Multiple tracing modes allow you to locate and trace energized and de-energized utilities in a variety of applications
  • The receiver’s high-contrast display allows for clear viewing in full sunlight and features an automatic backlight for shaded and dark areas
  • Semi-automatic gain control quickly detects tracing signal and allows precise adjustment of the receiver sensitivity
  • Depth measurement up to 20 ft
  • Rated CAT IV 600 V, ensuring safety when working with energized cables
  • Rugged, durable construction: water and dust resistant to IP54 and drop proof to 3.28 ft (1 m)
  • Use the Signal Clamp to induce a signal without making electrical contact (included with the UAT-620)
Accidentally hitting a utility line during a project can lead to costly repairs and create hazardous public safety situations. Digging in the wrong place can also lead to unnecessary delays and costs for your project, and ultimately, your company. Avoid this disruption with the rugged and durable Amprobe UAT-Series.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Amprobe UAT-505 Underground Utility Locator Kit WZ-20034-72 Mfr # Amprobe UAT-505
COMPARE
UAT-505 Underground Utility Locator Kit
Call For Price
Amprobe UAT-610 Underground Utility Locator Kit WZ-20034-73 Mfr # Amprobe UAT-610
COMPARE
UAT-610 Underground Utility Locator Kit
Call For Price
Amprobe UAT-620 Underground Utility Locator Kit with Signal Clamp WZ-20034-74 Mfr # Amprobe UAT-620
COMPARE
UAT-620 Underground Utility Locator Kit with Signal Clamp