This product is now discontinued, View Related Products
AEMC MiniFlex Flexible Current Probes
  • Measures from 0.5 Arms to 3000 Arms
  • Accuracy 1% of Reading ±0.2A
  • TRMS measurements when connected to a TRMS instrument
  • No core saturation or damage if overloaded
  • Overrange LED for measurement circuitry
  • EN 61010; 1000V CAT III; 600V CAT IV; CE Mark
  • 20kHz frequency response
  • Low phase shift for power measurements
  • Insensitive to DC, measures only AC component on DC + AC signals
  • Excellent linearity
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
AEMC 2126.81 Flexible Current Probe, 30/300A, 6 inch, 100mV/10mV/A WZ-20035-65 Mfr # 2126.81
COMPARE
Call For Price
AEMC 2126.82 Flexible Current Probe, 300/3000A, 10 inch, 10mV/1mV/A WZ-20035-66 Mfr # 2126.82