Advantec Disposable Syringe Filters, Nylon
Housings are specifically designed to minimize sample holdup and maximize recovery
  • Non-pigmented acrylic or polypropylene (PP) housings feature integral filter sealing to avoid the risk of contamination from pigments and adhesives
  • Each filter is clearly marked with membrane pore size
  • Housings have standard luer connectors; 50-mm filters feature 7 to 13.5 mm hose barb connectors
Nylon Filters are strong hydrophilic filters compatible with aqueous and alcoholic solutions and most HPLC solvents. They offer minimal extractables and high binding capacity for proteins, DNA, and RNA.
£193.71 - £363.22GBP / Pkg of 100
£232.45 - £435.86GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm)
Availability Pricing
Advantec 13NP045AN Syringe Filters, 0.45 um Nylon, 13 mm; 100/Pk WZ-81054-22 Mfr # 13NP045AN
COMPARE
13 0.45
£193.71GBP / Pkg of 100
£232.45GBP (incl VAT)
Advantec 30NP022AN Syringe Filters, 0.22 um Nylon, 30 mm; 100/Pk WZ-81054-52 Mfr # 30NP022AN
COMPARE
30 0.22
£363.22GBP / Pkg of 100
£435.86GBP (incl VAT)
Advantec 30NP045AN Syringe Filters, 0.45 um Nylon, 30 mm; 100/Pk WZ-81054-54 Mfr # 30NP045AN
COMPARE
30 0.45
£363.22GBP / Pkg of 100
£435.86GBP (incl VAT)