Advantec Air Sampling Filters, Quartz
Quartz filters ideal for sampling airborne particulates
Quartz Filters are used for air sampling of acidic gases at high temperatures. Quartz fibers are unaffected by humidity, highly resistant to chemical attack, and biologically inert. Filters are prefired at 1832°F (1000°C) for two hours to reduce organic contamination. Show very low trace metals content and do not absorb nitrous and sulfur dioxides.
£137.21 - £573.25GBP / Pkg of 100
£164.65 - £687.90GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec QR10047MM Air Sampling Filters, Quartz, 47-mm dia disc; pack of 100. WZ-06649-52 Mfr # QR10047MM
COMPARE
Air Sampling Filters, Quartz, 47-mm dia disc; pack of 100.
£137.21GBP / Pkg of 100
£164.65GBP (incl VAT)
Advantec QR1008X10IN Air Sampling Filters, Quartz, 8" x 10" (20 x 25 cm) sheet; pack of 50. WZ-06649-58 Mfr # QR1008X10IN
COMPARE
Air Sampling Filters, Quartz, 8\" x 10\" (20 x 25 cm) sheet; pack of 50.
£573.25GBP / Pkg of 50
£687.90GBP (incl VAT)