Adapter Fittings, Compression to Male Threaded, Straight
Excellent choice to connect rigid tubing systems
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
£7.06 - £46.63GBP / Each
£8.47 - £55.96GBP (incl VAT)
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size NPT(F) (port 2) (in)
Availability Pricing
COMPARE
1/8 PVDF
£7.06GBP / Each
£8.47GBP (incl VAT)
COMPARE
1/4 PVDF
£7.17GBP / Each
£8.60GBP (incl VAT)
COMPARE
1/4 PVDF
£8.66GBP / Each
£10.39GBP (incl VAT)
COMPARE
5/16 PVDF
£20.25GBP / Each
£24.30GBP (incl VAT)
COMPARE
3/8 PVDF
£11.08GBP / Each
£13.30GBP (incl VAT)
COMPARE
3/8 PVDF
£11.08GBP / Each
£13.30GBP (incl VAT)
COMPARE
1/2 PVDF
£30.93GBP / Each
£37.12GBP (incl VAT)
COMPARE
1/2 PVDF
£14.21GBP / Each
£17.05GBP (incl VAT)
COMPARE
5/8 PVDF
£31.52GBP / Each
£37.82GBP (incl VAT)
COMPARE
5/8 PVDF
£15.82GBP / Each
£18.98GBP (incl VAT)
COMPARE
3/4 PVDF
£46.63GBP / Each
£55.96GBP (incl VAT)
COMPARE
3/4 PVDF
£29.91GBP / Each
£35.89GBP (incl VAT)
COMPARE
1/16 Stainless Steel
£24.68GBP / Each
£29.62GBP (incl VAT)
Showing 1- 13 of 13