This product is now discontinued, View Related Products
Adam Equipment CQT Grain Scales
The CQT series grain scales have been developed specifically for agricultural use to calculate the potential yields and quality of the grain crop and it’s development.
  • 9 weighing units including pounds per bushel and grams or kilograms per hectoliter
  • Selectable settings for yield, dockage, and container size
  • Full range tare
  • Backlit LCD display
  • Stainless steel pan
  • ShockProtect™ three-point overload protection
  • Lock down mounting slot for Kensington™ type lock and cable
  • Dual tare keys
  • Removable anti-static draft shield
  • Lightweight and portable
  • Color coded, sealed keypad
  • External calibration
  • Auto sleep / power down function to save battery life
  • Auto power off
  • AC Adapter
  • Low battery indication
  • Below balance weighing with hanger
The CQT series grain scales are a simple, low cost solution that can be used in the field. The compact design and battery power operation makes transportation simple. Easily taking readings where the grain is located without the need of external power sources. The stainless steel pan can be removed for quick clean ups and the color coded keypad and smart functions makes operation trouble free.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment CQT 1752GR Grain Scale, 1750g Capacity and 0.05G Readability, 115V WZ-11810-00 Mfr # CQT 1752GR -115V
COMPARE
1.75 115
Call For Price
Adam Equipment CQT 1752GR Grain Scale, 1750g Capacity and 0.05G Readability, 220V WZ-11810-01 Mfr # CQT1752GR 220V
COMPARE
1.75 230