This product is now discontinued, View Related Products

Adam Equipment AFK Industrial Scales

IP66-rated stainless steel indicator is sealed from water and dust for tough environments
  • Extremely versatile indicator can be tilted, rotated 360°, mounted on the pillar, or attached to a wall
  • Battery and AC operation
  • Stainless steel indicator is sealed from water and dust
  • Adjustable pillar mounted indicator can be adjusted
  • Large backlit LCD display with capacity tracker
  • Parts counting with selectable sample size
  • Checkweighing LED lights show acceptable limits
  • Features six weighing units and five weighing modes
  • Removable stainless steel pan for easy cleaning
  • Automatic accumulation for adding totals
  • Bi-directional RS-232 output
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Adam Equipment AFK 165A Industrial Scale, 165Lb/75kg Capacity and 0.01Lb/5G Readability 115V WZ-11120-04 Mfr # AFK 165A-115V
COMPARE
75 165 115
Call For Price
Adam Equipment AFK 165A Industrial Scale, 165Lb/75kg Capacity and 0.01Lb/5G Readability 230V WZ-11120-06 Mfr # AFK 165A-230V
COMPARE
75 165 230
Call For Price
Adam Equipment AFK 330A Industrial Scale, 330lb/150K Capacity and 0.02Lb/10g Readability 115V WZ-11120-07 Mfr # AFK 330A-115V
COMPARE
150 330 115
Call For Price
Adam Equipment AFK 330A Industrial Scale, 330lb/150K Capacity and 0.02Lb/10g Readability 230V WZ-11120-08 Mfr # AFK 330A-230V
COMPARE
150 330 230
Call For Price
Adam Equipment AFK 660A Industrial Scale, 660lb/300K Capacity and 0.05Lb/20g Readability 115V WZ-11120-09 Mfr # AFK 660A-115V
COMPARE
300 660 115
Call For Price
Adam Equipment AFK 660A Industrial Scale, 660lb/300K Capacity and 0.05Lb/20g Readability 230V WZ-11120-11 Mfr # AFK 660A-230V
COMPARE
300 660 230
Call For Price
Adam Equipment AFK 1320A Industrial Scale, 1320lb Capacity and 0.1Lb/50g Readability 115V WZ-11120-14 Mfr # AFK 1320A-115V
COMPARE
600 1320 115
Call For Price
Adam Equipment AFK 1320A Industrial Scale, 1320lb Capacity and 0.1Lb/50g Readability 230V WZ-11120-16 Mfr # AFK 1320A-230V
COMPARE
600 1320 230
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat