Diba Activated Carbon Vapor Traps for T-Series Solvent Waste Caps
Prevent chemical vapors from escaping from solvent containers
  • Compatible with Diba T-series solvent caps
Use traps with Luer connection with T-series solvent waste caps without valve and filter. Traps with 1/4"-28 UNF(M) connection works with caps with vent. Traps with UNF connection feature an indicator strip that changes color as it gets saturated.
£60.33 - £1,257.50GBP / Each
£72.40 - £1,509.00GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Compatible With
Availability Pricing
Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Luer Ports; 1/ea WZ-21943-29 Mfr # 009CF-T
COMPARE
T-series solvent caps without valve and filter; male luer slip connection
£60.33GBP / Each
£72.40GBP (incl VAT)
Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Venting Port; 1/ea WZ-21943-30 Mfr # 009CF-TW
COMPARE
T-series solvent caps with vent; 1/4\"-28 UNF(M) connection
£260.75GBP / Each
£312.90GBP (incl VAT)
Diba Omnifit® Activated Carbon Vapor Trap for Solvent Waste Caps with Venting Port; 5/pk WZ-21943-31 Mfr # 009CF-TWB
COMPARE
T-series solvent caps with vent; 1/4\"-28 UNF(M) connection
£1,257.50GBP / Pkg of 5
£1,509.00GBP (incl VAT)