AccuSense High Accuracy Differential Pressure Transducers
Rugged stainless steel construction provides reliability in demanding applications
  • High accuracy of ±0.07% full-scale
  • NEMA 4X/IP65 rated
  • Very low thermal error
Designed for use with air and clean gases, Setra''s AccuSense ASL series of transducers are designed for low differential pressure measurement where accuracy and durability are required. Although the tranducers have a very high accuracy measurement of low pressures, they have a high proof pressure capacity. Transducers can be used for numerous types of applications where diferential pressure measurment is required, such as test stands, leak detection, and pharmaceutical manufacuring, but the robust design and construction makes these perfect for industrial manufacuring and process applications. Choose from positive or bidirectional differential ranges. Transducer comes with metal mounting bracket. Other engineering units are available — contact an application specialist for details.
Input and output is wired via male 6-pin bayonet connector 98075-90. Order SecureCal™ calibrator 98075-95 to calibrate tansducers in the field—enables zero-tare and snap-span adjustment, factory calibration restore function, and two-line LCD and LED feedback.
£2,500.00GBP / Each
£3,000.00GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (in WC) Min Pressure (in WC)
Availability Pricing
Setra ASL Bidirectional Differential Pressure Transmitters +/-5 in WC WZ-98075-84 Mfr # ASL1-005WB-1F-2C-B3-A-01
COMPARE
±0.07% full-scale 5 -5
£2,500.00GBP / Each
£3,000.00GBP (incl VAT)