accumet AB Series Benchtop Meters
Combines basic meter economy with 0.001 pH resolution
  • Plain language prompts for easy setup
  • 500 data points with date-and-time stamp to meet GLP
  • USB and bidirectional RS-232 connectivity
The accumet AB-series meters offer benefits typically found on more expensive meters — a graphic LCD, bidirectional RS-232 and USB connectivity, password protection, and GLP compliant 500 datapoint memory. The large, backlit display lets you easily view measurements with calibration data; plus a graphic electrode status indicator allows you to verify electrode condition at a glance.
The meter accepts up to five-point pH calibration with custom or preset buffer groups in pH mode. It also offers calibration and high/low alarms for system monitoring. ISE mode offers direct measurements in ppm, mg/L, and molar concentration. Meter accepts pH and ISE electrodes with a BNC connector, and two- and four-cell conductivity probes with cell constants equal to 0.1, 1.0, and 10.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Accumet AB150 pH Benchtop Meter Only WZ-59331-52 Mfr # 13636AB150A
COMPARE
pH Benchtop Meter Only
Call For Price
Accumet AB250 pH/ISE Benchtop Meter Only WZ-59331-54 Mfr # 13636AB250A
COMPARE
pH/ISE Benchtop Meter Only
Call For Price
Accumet AB200 pH/Conductivity Benchtop Meter Only WZ-59331-56 Mfr # 13636AB200A
COMPARE
pH/Conductivity Benchtop Meter Only
Call For Price
Fisher Accumet AB150 pH Benchtop Meter Kit WZ-59340-00 Mfr # 13636AB150
COMPARE
pH Benchtop Meter Kit
Call For Price
Fisher Accumet AB150B pH Benchtop Meter Kit WZ-59340-05 Mfr # 13636AB150B
COMPARE
pH Benchtop Meter Kit
Call For Price
Fisher Accumet AB200 pH / ISE Benchtop Meter Kit WZ-59340-10 Mfr # 13636AB200
COMPARE
pH / ISE Benchtop Meter Kit
Call For Price
Fisher Accumet AB250 pH/Conductivity Benchtop Meter Kit WZ-59340-20 Mfr # 13636AB250
COMPARE
pH/Conductivity Benchtop Meter Kit
Call For Price
Showing 1- 7 of 7