This product is now discontinued, View Related Products

Accident Prevention Tags; Danger: Do Not Operate

OSHA/ANSI-Compliant Header Tags meet OSHA 29 CFR 1910.145 (f) and ANSI Z535.5 standards which provide specifications, designs, and guidelines for the use and application of accident prevention tags. These tags clearly indicate a temporary hazard that may lead to personal injury or property damage; use until that hazard is eliminated or hazardous operation is complete.

Choose economical cardstock which is a 10-mil polyolefin based material containing no wood-based paper pulp, or durable, rigid 15-mil vinyl plastic material which withstands tears, weathering, and various chemicals.

All tags can be written on and have top gabled corners with 3⁄8" reinforced metal grommet on vinyl plastic.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Legend Material
Availability Pricing
Accuform MDT112CTP Tag, Danger Do Not Operate, Back A, 5 7/8" X 3 1/8", PF-Cardstock WZ-40503-48 Mfr # MDT112CTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! TO DO SO WITHOUT AUTHORITY WILL MEAN DISCIPLINARY ACTION! IT IS HERE FOR A PURPOSE. Cardstock
Call For Price
Accuform MDT112PTP Tag, Danger Do Not Operate, Back A, 5 7/8" X 3 3/8", RV-Plastic WZ-40503-49 Mfr # MDT112PTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! TO DO SO WITHOUT AUTHORITY WILL MEAN DISCIPLINARY ACTION! IT IS HERE FOR A PURPOSE. Plastic
Call For Price
Accuform MDT189CTP Tag, Danger Do Not Operate, Back B, 5 7/8" X 3 1/8", PF-Cardstock WZ-40503-50 Mfr # MDT189CTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! Cardstock
Call For Price
Accuform MDT189PTP Tag, Danger Do Not Operate, Back B, 5 7/8" X 3 3/8", RV-Plastic WZ-40503-51 Mfr # MDT189PTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! Plastic
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat