This product is now discontinued, View Related Products
Accessories for Koehler Existent Gum Test Apparatus
Optimize performance of your Koehler Existent Gum Evaporation Bath
Compatible for use with the Koehler Existent Gum Evaporation Bath model K33700. Bath sold separately.
Steam Superheater provides dry superheated steam for evaporation baths not equipped with a built-in superheater. Use together with an outside steam source for steam-jet method testing of aircraft turbine fuels. Superheating chamber and condensate trap are constructed entirely of stainless steel. Solid state temperature controller adjusts between 0-550°F. Equipped with steam inlet and outlet connections and condensate drain valve. Steel exterior has a chemical resistant polyurethane enamel finish.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Koehler K33850/208603 Steam Boiler, 208 VAC, 60 Hz, Three-Phase WZ-59876-13 Mfr # K33850/208603
COMPARE
Steam Boiler, 208 VAC, 60 Hz, Three-Phase
Call For Price
Koehler K33810 Steam Superheater, 220-240 VAC, 50/60 Hz WZ-59876-14 Mfr # K33810
COMPARE
Steam Superheater, 220-240 VAC, 50/60 Hz