Absorptive Silencer for Compressors or Vacuum Pumps
Air intake and discharge filter silencers offer minimal pressure drop
  • Layered sound-absorbent media
  • Minimal pressure drop because it does not rely on internal baffles, tubes or other restrictive devices
  • Reduces high frequency noise to 30 dB
  • Inlet or discharge silencing applications with maximum temperature of 212°F (100°C)
  • Durable in-line carbon steel construction with baked-enamel finish
Ideal for:
Small air compressors
Centrifugal blowers
Regenerative blowers
Vacuum pumps and systems
£126.35 - £157.87GBP / Each
£151.62 - £189.44GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Description
Availability Pricing
COMPARE
42 Absorptive Silencer for Compressors or Vacuum Pumps, 42 SCFM; 12\" L x 2.5\" dia
£126.35GBP / Each
£151.62GBP (incl VAT)
COMPARE
155 Absorptive Silencer for Compressors or Vacuum Pumps, 155 SCFM; 12\" L x 3.125\" dia
£157.87GBP / Each
£189.44GBP (incl VAT)