Abaque™ Peristaltic Natural Rubber Replacement Hose
Highly resilient with excellent abrasion resistance and strength.
  • All listed hoses in this ad are made from Natural Rubber.
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Abaque engineers / manufactures their hoses from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression.
Ideal for applications where the following properties are needed:
Highly resilient, excellent abrasion resistance, the most common hose in peristaltic markets. Generally resistant with lightly corrosive chemical applications, abrasive slurries, etc.
£115.50 - £2,377.20GBP / Each
£138.60 - £2,852.64GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (mm) Tubing OD (mm)
Availability Pricing
Abaque Y218712 HDN10 Replacement Hose - Natural Rubber WZ-75810-21 Mfr # Y218712
COMPARE
1.67 10 32
£115.50GBP / Each
£138.60GBP (incl VAT)
Abaque Y218938 HDN15 Replacement Hose - Natural Rubber WZ-75810-26 Mfr # Y218938
COMPARE
2.47 15 37
£163.80GBP / Each
£196.56GBP (incl VAT)
Abaque Y218941 HDN20 Replacement Hose - Natural Rubber WZ-75810-32 Mfr # Y218941
COMPARE
2.47 20 37
£215.25GBP / Each
£258.30GBP (incl VAT)
Abaque Y218721 HDN25 Replacement Hose - Natural Rubber WZ-75810-37 Mfr # Y218721
COMPARE
3.28 25 54
£267.75GBP / Each
£321.30GBP (incl VAT)
Abaque Y218724 HDN32 Replacement Hose - Natural Rubber WZ-75810-43 Mfr # Y218724
COMPARE
4.1 32 63
£354.90GBP / Each
£425.88GBP (incl VAT)
Abaque Y218727 HDN40 Replacement Hose - Natural Rubber WZ-75810-48 Mfr # Y218727
COMPARE
3.28 38 63
£414.75GBP / Each
£497.70GBP (incl VAT)
Abaque Y218730 HDN40X Replacement Hose - Natural Rubber WZ-75810-54 Mfr # Y218730
COMPARE
4.1 40 63
£466.20GBP / Each
£559.44GBP (incl VAT)
Abaque Y218733 HDN50 Replacement Hose - Natural Rubber WZ-75810-59 Mfr # Y218733
COMPARE
5.97 50 80
£604.80GBP / Each
£725.76GBP (incl VAT)
Abaque Y218736 HDN65 Replacement Hose - Natural Rubber WZ-75810-64 Mfr # Y218736
COMPARE
5.97 60 87
£983.85GBP / Each
£1,180.62GBP (incl VAT)
Abaque Y218739 HDN65X Replacement Hose - Natural Rubber WZ-75810-69 Mfr # Y218739
COMPARE
7.66 65 100
£1,009.05GBP / Each
£1,210.86GBP (incl VAT)
Abaque Y218742 HDN80X Replacement Hose - Natural Rubber WZ-75810-73 Mfr # Y218742
COMPARE
7.66 80 115
£1,239.00GBP / Each
£1,486.80GBP (incl VAT)
Abaque Y218745 HDN80 Replacement Hose - Natural Rubber WZ-75810-78 Mfr # Y218745
COMPARE
9.12 80 123
£1,587.60GBP / Each
£1,905.12GBP (incl VAT)
Abaque Y218748 HDN100 Replacement Hose - Natural Rubber WZ-75810-83 Mfr # Y218748
COMPARE
10.74 100 144
£2,377.20GBP / Each
£2,852.64GBP (incl VAT)
Abaque 130083 15mm Natural Rubber Custom Replacement Hose WZ-75810-97 Mfr # 130083
COMPARE
£214.20GBP / Each
£257.04GBP (incl VAT)
Abaque 130088 25mm Natural Rubber Custom Replacement Hose WZ-75811-02 Mfr # 130088
COMPARE
£348.60GBP / Each
£418.32GBP (incl VAT)
Abaque 130093 32mm Natural Rubber Custom Replacement Hose WZ-75811-07 Mfr # 130093
COMPARE
£462.00GBP / Each
£554.40GBP (incl VAT)
Abaque 130098 40Xmm Natural Rubber Custom Replacement Hose WZ-75811-12 Mfr # 130098
COMPARE
£607.95GBP / Each
£729.54GBP (incl VAT)
Abaque 130103 50mm Natural Rubber Custom Replacement Hose WZ-75811-17 Mfr # 130103
COMPARE
£786.45GBP / Each
£943.74GBP (incl VAT)
Showing 1- 18 of 18