Abaque™ Buna-N (FDA liner) Replacement Hose
For hygienic applications (FDA approved)
  • All listed hoses in this ad are made from Buna-N (FDA liner).
  • The hose IDs are: 10, 15, 20, 25, 32, 40, 50, 65, 80, 100 mm. Each liquid end is designed to accept one particular hose ID version.
The hose of a Peristaltic Hose pump is the only component that comes in contact with the pumped medium. Neptune engineers hoses the are manufactured from high quality compounded rubber, reinforced by layers of nylon. Once constructed, the hose is then machined with precision for efficiency, accuracy, longevity, and optimal compression.
£229.95 - £2,607.15GBP / Each
£275.94 - £3,128.58GBP (incl VAT)
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (mm) Tubing OD (mm)
Availability Pricing
Abaque Y229474 HDN10 Replacement Hose - Buna-N (FDA liner) WZ-75810-23 Mfr # Y229474
COMPARE
1.67 10 32
£229.95GBP / Each
£275.94GBP (incl VAT)
Abaque Y229475 HDN15 Replacement Hose - Buna-N (FDA liner) WZ-75810-28 Mfr # Y229475
COMPARE
2.47 15 37
£340.20GBP / Each
£408.24GBP (incl VAT)
Abaque Y229154 HDN20 Replacement Hose - Buna-N (FDA liner) WZ-75810-34 Mfr # Y229154
COMPARE
2.47 20 37
£374.85GBP / Each
£449.82GBP (incl VAT)
Abaque Y229476 HDN25 Replacement Hose - Buna-N (FDA liner) WZ-75810-39 Mfr # Y229476
COMPARE
3.28 25 54
£430.50GBP / Each
£516.60GBP (incl VAT)
Abaque Y229482 HDN32 Replacement Hose - Buna-N (FDA liner) WZ-75810-45 Mfr # Y229482
COMPARE
4.1 32 63
£465.15GBP / Each
£558.18GBP (incl VAT)
Abaque Y229380 HDN40 Replacement Hose - Buna-N (FDA liner) WZ-75810-51 Mfr # Y229380
COMPARE
3.28 38 63
£467.25GBP / Each
£560.70GBP (incl VAT)
Abaque Y229477 HDN40X Replacement Hose - Buna-N (FDA liner) WZ-75810-56 Mfr # Y229477
COMPARE
4.1 40 63
£485.10GBP / Each
£582.12GBP (incl VAT)
Abaque Y229483 HDN50 Replacement Hose - Buna-N (FDA liner) WZ-75810-61 Mfr # Y229483
COMPARE
5.97 50 80
£1,059.45GBP / Each
£1,271.34GBP (incl VAT)
Abaque Y229155 HDN65 Replacement Hose - Buna-N (FDA liner) WZ-75810-66 Mfr # Y229155
COMPARE
5.97 60 87
£1,228.50GBP / Each
£1,474.20GBP (incl VAT)
Abaque Y229484 HDN65X Replacement Hose - Buna-N (FDA liner) WZ-75810-70 Mfr # Y229484
COMPARE
7.66 65 100
£1,168.65GBP / Each
£1,402.38GBP (incl VAT)
Abaque Y229479 HDN80X Replacement Hose - Buna-N (FDA liner) WZ-75810-75 Mfr # Y229479
COMPARE
7.66 80 115
£1,429.05GBP / Each
£1,714.86GBP (incl VAT)
Abaque Y229478 HDN80 Replacement Hose - Buna-N (FDA liner) WZ-75810-80 Mfr # Y229478
COMPARE
9.12 80 123
£1,647.45GBP / Each
£1,976.94GBP (incl VAT)
Abaque Y229480 HDN100 Replacement Hose - Buna-N (FDA liner) WZ-75810-85 Mfr # Y229480
COMPARE
10.74 100 144
£2,607.15GBP / Each
£3,128.58GBP (incl VAT)
Abaque 130085 15mm Buna-N (FDA Liner) Custom Replacement Hose WZ-75810-99 Mfr # 130085
COMPARE
£442.05GBP / Each
£530.46GBP (incl VAT)
Abaque 130090 25mm Buna-N (FDA Liner) Custom Replacement Hose WZ-75811-04 Mfr # 130090
COMPARE
£558.60GBP / Each
£670.32GBP (incl VAT)
Abaque 130095 32mm Buna-N (FDA Liner) Custom Replacement Hose WZ-75811-09 Mfr # 130095
COMPARE
£604.80GBP / Each
£725.76GBP (incl VAT)
Abaque 130100 40Xmm Buna-N (FDA Liner) Custom Replacement Hose WZ-75811-14 Mfr # 130100
COMPARE
£631.05GBP / Each
£757.26GBP (incl VAT)
Abaque 130105 50mm Buna-N (FDA Liner) Custom Replacement Hose WZ-75811-19 Mfr # 130105
COMPARE
£1,378.65GBP / Each
£1,654.38GBP (incl VAT)
Showing 1- 18 of 18