A&D Weighing SW Washdown Industrial Scales
IP69K–rated—the ultimate protection against the effects of water and dust
  • Stainless steel weighing pan, frame, and load cell
  • Designed to withstand heavy-duty cleaning at temperatures up to 175°F and pressures up to 1450 psi
  • Bright, multicolored display with 5-step comparator and touch-sensitive capacitive keys
  • Attach the indicator pole to either the short or long side of the platform
  • Indicator bracket features an adjustable bracket for convenient viewing angles
  • Counting and checkweighing
£2,059.22GBP / Each
£2,471.06GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing SW-6KS Washdown Industrial Scale, Ssip69K, 13Lb/6kg, NSF; 9.75" WZ-11110-67 Mfr # SW-6KS
COMPARE
6 13 100 to 240
£2,059.22GBP / Each
£2,471.06GBP (incl VAT)