A&D Weighing HW-G and HV-G Series Industrial Scales
Tilting LCD feature enhances visibility
  • Comparator for setting high and low weight targets
  • Tilting meter head for best viewing angle
  • Counting and percent function
These scales provide counting, percentage, and comparator functions. Ideal for packaging, shipping, pharmaceutical, and industrial applications.
HV-G series of scales offer resolution that can be changed automaticlly for 3 Capacity levels.
HW-G series of scales has high resolution of 1/10,000.
V model scales have a Vacuum Fluorescent display, that is ideal for use in dark areas.
L model scales have a LCD display.
£630.00 - £956.00GBP / Each
£756.00 - £1,147.20GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing HV-15KGL Triple Resolution Bench Scale, LCD;  6/15/30 lb WZ-11122-04 Mfr # HV-15KGL
COMPARE
15 115 / 230
£630.00GBP / Each
£756.00GBP (incl VAT)
A&D Weighing HV-200KGL Triple Resolution Bench Scale, LCD, 150/300/500 lb SKU 1112210 WZ-11122-10 Mfr # HV-200KGL
COMPARE
500 115 / 230
£762.00GBP / Each
£914.40GBP (incl VAT)
A&D Weighing HW-10KGL High Resolution Bench Scale with LCD; 20 lb x 0.002 lb WZ-11125-00 Mfr # HW-10KGL
COMPARE
10 115
£883.00GBP / Each
£1,059.60GBP (incl VAT)
A&D Weighing HW-100KGL High Resolution Bench Scale with LCD; 200 lb x 0.02 lb WZ-11125-20 Mfr # HW-100KGL
COMPARE
100 200 115 / 230
£952.00GBP / Each
£1,142.40GBP (incl VAT)
A&D Weighing HW-200KGL High Resolution Bench Scale with LCD; 500 lb x 0.05 lb WZ-11125-30 Mfr # HW-200KGL
COMPARE
220 500 115 / 230
£956.00GBP / Each
£1,147.20GBP (incl VAT)