This product is now discontinued, View Related Products
A&D PV Handheld Balances
Offers high performance at a low price
  • Protective vinyl cover keeps dust out
  • Low battery indicator
  • Multiple weighing units (g, oz, dwt, ozt)
The compact, portable balances feature fast and simple calibration, battery operation, and auto-power off to save battery life. Ideal for jewelry weighing, hobby applications, manufacturing, quality control, packaging, sample preperation, field testing, checkweighing, environmental testing, postal scale, and more.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Battery Capacity (g) Readability (g)
Availability Pricing
A&D Weighing PV-60 PV Handheld Balance, 60g x 0.01g - +/- 0.02g WZ-11111-40 Mfr # PV-60
COMPARE
60 0.01
Call For Price
A&D Weighing PV-100 Handheld Balance, 100g x 0.1g -+/- 0.2g WZ-11111-42 Mfr # PV-100
COMPARE
2 x AAA (included) 100 0.1
Call For Price
A&D Weighing PV-200 Handheld Balance, 200g x 0.1g -+/- 0.2g WZ-11111-44 Mfr # PV-200
COMPARE
2 x AAA (included) 200 0.1
Call For Price
A&D Weighing PV-500 Handheld Balance, 500g x 0.1g -+/- 0.2g WZ-11111-46 Mfr # PV-500
COMPARE
2 x AAA (included) 500 0.1