Costar® 96-Well Microplate Storage Mat
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Manufactured from EVA (ethyl vinyl acetate) and polymer is compatible with DMSO
  • Can be applied manually or with our Storage Mat Applicator
Storage mat seals each well individually for long-term storage—virtually eliminates moisture loss. Use storage mats with standard microplates and 1.0-mL deep-well assay blocks.
£449.08GBP / Case of 100
£538.90GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Material Surface Treatment Description
Availability Pricing
Costar Microplate Storage Mat, 96-Well, EVA; 100/Cs WZ-01729-22 Mfr # 3080
COMPARE
Ethyl Vinyl Acetate Not Treated Microplate Storage Mat, 96-Well, EVA; 100/Cs
£449.08GBP / Case of 100
£538.90GBP (incl VAT)