Thermo Scientific™ SureSTART™ Certified Shell Vial and Plug Kits
Vial kits combine vials and plugs for your convenience
  • Certified performance level 1 for everyday analysis and chromatography approved
  • Constructed of glass vials and polyethylene plugs
Meet your certification and documentation needs with the Level 1 Thermo Scientific™ SureSTART™ specification certified shell vial and plug kits. These convenience kits contain 1.25 mL shell screw vials in clear glass and transparent 8 mm polyethylene plug closures.
Level 1 products are cost optimized, suitable for all instrument types, and quality tested to deliver the performance, sample security, and reproducibility you need for everyday analyses. They are an ideal solution for verifying compound presence and quantification using detection techniques such as UV, RI, and FID.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Type Level Volume (mL)
Availability Pricing
Thermo Scientific SureSTART™ Certified Shell Vial and Plug Kit, Level 1, Clear Glass, 1.25 mL; 200/Pk WZ-08922-47 Mfr # 6EKSHV8-1
COMPARE
Plug 1 1.25