Masterflex® Ismatec® Microflow Series Peristaltic Pumps
With flow ranges from 0.0001 to 365 mL/min, Masterflex Ismatec microflow pumps with low flow that gives you precise control. Ismatec microflow pumps are ideal for autoanalysis, flow cell, sipper, OEM, and low flow feed and transfer applications. They offer the latest technology including MasterflexLive, easy control and tubing changes, and the low-maintenance and cost-effectiveness of peristaltic pumps.
Ismatec pumps accept Ismatec pump tubing for clean fluid paths. Tubing changes are quick and easy with several available tubing formulations to ensure compatibility with your fluid.
MORE +
Ismatec is now part of the Masterflex Brand of precision peristaltic pumps
Featured Products