This item is now discontinued. Please browse similar items
ZenPure Hepa capsule filter, 625 cm2 filtration area
Representative image only.
ZenPure Mfr # SKLPHH2H2H – Item # WZ-49700-92
Ideal for pharmaceutical, biotechnology, and food and beverage applications.
  • 99.97% efficiency at 0.3 µm
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialHEPA
  • Housing MaterialPolypropylene
  • Connections1/4\" to 1/2\" hose barb
  • Effective Filtration Area (cm²)600
  • Surface Area (m²)0.06
  • Particle Retention (µm)0.3
  • Particle Retention (%)99.97
  • DescriptionHepa capsule filter, 625 cm2 filtration area
Key Features
  • 99.97% efficiency at 0.3 µm
More About this Item
Designed to provide bacteria-, algae-, fungal-free air for sterile applications, such as fermentors or incubators. The media allows for high flow rates and low pressure drop. Designed for high retention and particle holding capacity for air and gas. Housing is made of durable polypropylene.