YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 30 m Cable
Representative image only.
YSI Mfr # 627150-30 – Item # WZ-59353-82
Get lab-quality results at the source
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 30-m cable
  • 2 year warranty
PRODUCT OPTIONS
£1,430.96GBP / Each
£1,717.15GBP (incl VAT)
Include Meter Today!
YSI ProSolo Handheld Optical Dissolved Oxygen Meter Only
£881.04GBP / Each
£1,057.25GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsWater quality, waste water, natural resources, oxygen analysis
  • Min Temperature (° C)-5
  • Max Temperature (° C)70
  • Temperature CompensationYes
  • GuardStainless steel
  • ConnectorWaterproof
  • Cable Length (m)30
  • DescriptionOptical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 30 m Cable
Two year warranty
Includes
Probes with cables 13-ft (4-m) to 100-ft (30-m) include cable management kit.
Key Features
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 30-m cable
  • 2 year warranty
More About this Item
Interchangeable digital "smart" probes store calibration data for a year, saving you time and calibration solution costs—no need to recalibrate if connected to different meters. Convenient cable management kits are included with probes featuring cable lengths from 13-ft (4-m) to 100-ft (30-m)