This item is now discontinued. View the direct replacement: WZ-35604-10 or browse similar items
YSI EC300 Ecosense Conductivity Meter, without Probes
Representative image only.
YSI Mfr # 606079 – Item # WZ-19750-30
Accurate, economical portable meter with four-electrode conductivity cell
  • Rugged, weighted probe assembly with automatic temperature compensation
  • Measures conductivity, salinity or TDS with temperature
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypePortable
  • Display typeLCD
  • Min Conductivity0.0 to 200 mS
  • Conductivity Resolution0.1 μS or 0.1 mS
  • Conductivity Accuracy±1% of reading
  • Salinity Resolution0.1 ppt
  • Temperature compensationAutomatic or manual
  • Data Logging (points)50 data points
  • DescriptionEcosense Conductivity Meter, without Probes
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 67
Key Features
  • Rugged, weighted probe assembly with automatic temperature compensation
  • Measures conductivity, salinity or TDS with temperature
  • 1 year warranty
More About this Item
The EC300A features easy-to-use interface, one-hand operation, and waterproof housing with rugged, weighted probes that can be ordered with 13- or 33-ft (4- or 10-m) cable. The rugged, four-electrode conductivity cell provides stable measurements from 0 to 200 mS with auto-ranging function that eliminates the need for multiple conductivity cell constants. Meter also offers auto shutoff, low battery indicator, and nonvolatile 50 data sets memory with calibration data.
Purchase meter with probes separately to customize your system, or choose a kit for a complete system.
Order probes separately.