SI Analytics BlueLine 11 pH, Glass pH Electrode with Plug Head
Representative image only.
Mfr # 285129114(BLUE LINE#11PH) – Item # WZ-55503-00
No more corrosion and interference from Ag/AgCI ions
  • Silamid® reference system eliminates corrosion and interference from silver/silver chloride ions
  • Platinum diaphragm and free flow of electrolyte ensure excellent stability
  • 1 year warranty
PRODUCT OPTIONS
£241.46GBP / Each
£289.75GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference JunctionSilimid reference system
  • Min pH (pH)0
  • Max pH (pH)14
  • Max Temperature (° C)100
  • ConnectorDepends upon cable purchased separately: BNC or DIN
  • DescriptionBlueLine 11 pH, Glass pH Electrode with Removeable Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Silamid® reference system eliminates corrosion and interference from silver/silver chloride ions
  • Platinum diaphragm and free flow of electrolyte ensure excellent stability
  • 1 year warranty
More About this Item
Laboratory work has never been easier with these new high-quality electrodes from SI Analytics. Innovative design features a sliding gate that allows easy filling of electrolyte solution and seals it from the solution being measured. BlueLine electrodes feature fast adjustment of measured values and high mechanical stability. Operating range is 0 to 14 pH.
Electrode models 55503-00, -30, and -46 offer superior durability with a cone-shaped electrode tip that is rugged and easy to clean. BNC and DIN fixed cable electrodes include a 3-ft (1-m) cable. Order cable 55503-50 with DIN connector or 55503-55 with BNC connector (sold separately) for use with electrodes with removable cable
ATC element is NTC 30 kΩ which is compatible with WTW meters
Schott—Reg TM Schott Glas