This item is now discontinued. Please browse similar items

WTW PH 7110 inoLab 7110 basic pH/mV/T benchtop meter

Representative image only.
WTW Mfr # 1AA120 – Item # WZ-58890-60
Providing accurate measurements, compliant documentation, and secure traceability
  • Auto-recognition of USA/NIST and DIN buffers
  • AutoRead function ensures repeatable results
  • Easy to calibrate and maintain with calibration timer
  • Power with universal AC adapter or AA batteries
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.005 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 3 points
  • Min mV (mV)-1200
  • Max mV (mV)1200
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.3 mV + 1 digit
  • Min Temperature (° C)-5
  • Max Temperature (° C)105
  • Min Temperature (° F)23
  • Max Temperature (° F)221
  • WaterproofYes
  • Power (VAC)100 to 240
  • Power (Hz)50 or 60
  • Battery4 x AA batteries (optional)
  • Width (cm)7.9375
  • Height (cm)19.05
  • Width (in)3 1/8
  • Length (in)9 1/2
  • Height (in)7 1/2
  • DescriptioninoLab 7110 basic pH/mV/T benchtop meter
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP54 Rated

Key Features

  • Auto-recognition of USA/NIST and DIN buffers
  • AutoRead function ensures repeatable results
  • Easy to calibrate and maintain with calibration timer
  • Power with universal AC adapter or AA batteries
  • 3 year warranty

More About this Item

The inoLab family of benchtop meters offer an intuitive user interface, large informative display, and keypad with tactile feedback to provide an exceptional user experience for error-free operation.

inoLab pH 7110 Basic Meter is ideally suited for routine, general laboratory pH/mV measurements. An intuitive user interface and easy-to-clean housing make it ideal for any environment.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat