WTW FDO 925-3 IDS Dissolved Oxygen Probe; 3 m Cable

Representative image only.
WTW Mfr # G201301 – Item # WZ-99575-43
  • 3 year warranty

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • Accessory TypeOptical DO Probe
  • Range 0 to 20.00 mg/L (ppm) at ± 0.5 % of value
  • MaterialPlastic and SS
  • DescriptionIDS Dissolved Oxygen Probe; 3 m Cable
  • Warranty3 year
3 Year WarrantyConformite Europeenne

Key Features

  • 3 year warranty

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat