This item is now discontinued. Please browse similar items
WTW 285126511 Conductivity Standard, 1.41 mS (1413 uS); 18 Ampoules
Representative image only.
WTW Mfr # 285126511 – Item # WZ-59351-44
Offering up to 5 years shelf life
  • Highly precise standards with ±0.25% accuracy
  • Certified accuracy traceable to NIST/DIN standards
  • Single-use buffers with 25-mL of solution
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)25
  • Volume (oz)0.9
  • Volume18 Ampoules
  • DescriptionConductivity Standard, 1.41 mS (1413 uS); 18 Ampoules
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Highly precise standards with ±0.25% accuracy
  • Certified accuracy traceable to NIST/DIN standards
  • Single-use buffers with 25-mL of solution
  • 2 year warranty
More About this Item
Presterilized and free of preservatives, these ready-to-use, sealed glass ampoules of conductivity standards offer a 5-year shelf life. 25 mL solution per ampoule.