Worthington R05K-8C00 Roller Base for Cryogenic Storage Dewars
Representative image only.
Worthington Mfr # R05K-8C00 – Item # WZ-03778-75
  • 3 month warranty
PRODUCT OPTIONS
£567.53GBP / Each
£681.04GBP (incl VAT)
often bought with
Traceable® Cryogenic Wi-Fi Data Loggers Compatible with TraceableLIVE® Cloud Service
£433.34 - £455.48GBP / Each
£520.01 - £546.58GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRoller Base for Cryogenic Storage Dewars
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Easily move storage Dewars and Dewars with canister storage systems. Sturdy cast aluminum frame has five wheels with ball-bearing swivel casters. Use model 03773-65 for Dewars with 15.6" diameter base; use 03773-71 for Dewars with 18.8" diameter base. Shpg wt 8.0 lb (3.7 kg). Note: Roller base does not include tipping stand.