Winters Instruments Q90211 Quality Process Gauge, 0-15 psi - 2 1/2" dial
Representative image only.
Winters Instruments Mfr # P9S90211 – Item # WZ-68612-02
Flutterless movement gives filled-like readings at lower cost
  • Screw-on lens makes the gauge easy to service and calibrate without removing gauge from process line
  • 3 year warranty
£23.19GBP / Each
£27.83GBP (incl VAT)
£159.35 - £197.25GBP / Each
£191.22 - £236.70GBP (incl VAT)
for an additional
£159.35GBP / Each
£191.22GBP (incl VAT)
for an additional
£197.25GBP / Each
£236.70GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2-1-2% (ANSI/ASME Grade B)
  • Dial Size (in)2.5
  • Dial Size (mm)64
  • Min Pressure (PSI)0
  • Max Pressure (PSI)15
  • Wetted Materialsbronze
  • Casesteel
  • Lens Materialacrylic
  • Process Connection1/4\" NPT(M)
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionQuality Process Gauge, 0-15 psi - 2 1/2\" dial
  • Warranty3 year
3 Year Warranty
Key Features
  • Screw-on lens makes the gauge easy to service and calibrate without removing gauge from process line
  • 3 year warranty
More About this Item
Process gauges are versatile instruments that come standard with a screw-on acrylic lens and recalibrator screw. Pressure gauge is also equipped with a flutterless movement for added stability. Gauges have phosphor bronze Bourdon tube, brass socket, black steel case, and polycarbonate lens.