Wildco 12-C82 Surber Stream Bottom Sampler, 1000 um net
Representative image only.
Wildco Mfr # 12-C82 – Item # WZ-05491-18
Quantitative sampler for benthic invertebrates
  • Recommended for sampling shallow streams
  • Open base encloses area of 12 x 12"
  • 3 month warranty
PRODUCT OPTIONS
£400.51GBP / Each
£480.61GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel, Nitex Nylon
  • DescriptionSurber Stream Bottom Sampler, 1000 um net
  • Warranty3 month
3 Month Warranty
Key Features
  • Recommended for sampling shallow streams
  • Open base encloses area of 12 x 12"
  • 3 month warranty
More About this Item
Also known as a riffle or run sampler this sampler has two pivoting stainless steel frames with lockable braces to hold the frames at right angles to each other. The net is 29" long with a filter area that is 17.5" long