This item is now discontinued. Please browse similar items
Wika GAUGE 6IN 5000PSI 1/2 B FF
Representative image only.
Wika Mfr # 4214421-0001 – Item # WZ-68433-56
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These gauges feature 316 stainless steel wetted parts and 304 stainless steel case and bayonet ring, an adjustable pointer, a laminated safety glass window, and can be liquid-filled in the field.
WIKA stainless steel gauges are ideal for process, chemical, and petrochemical applications, oil exploration and production, power generation, pollution control equipment, and any other application requiring fine quality, long lasting pressure measurement instrumentation.
  • Center Back Mount Connection Available
  • 1/4" NPT Connection Size
  • 4" Dial Size Available