This item is now discontinued. Please browse similar items
Wika BIMETAL 3IN 0/250F 4IN B
Representative image only.
Wika Mfr # 30040A206G4 – Item # WZ-90423-36
SPECS & DESCRIPTION
Specifications & Description
More About this Item
Our Process Grade bimetal thermometers offer a 3" and 5" dial size
and are ideal for most rugged industrial temperature measurement
applications. The hermetically-sealed case offers protection from
weather and dust, and is guaranteed against fogging up. An inert gel
is used to minimize pointer oscillation. These thermometers are guaranteed for seven years.
WIKA Bimetal thermometers are manufactured to ASME B40.3
standards.