This item is now discontinued. Please browse similar items
Wika 32025D012G4 Bimetal Thermometer 3" 200/1000F 2.5" L, 1/2" NPT Adjustable Angle
Representative image only.
Wika Mfr # 32025D012G4 – Item # WZ-90552-81
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)200
  • Max Temperature (° F)1000
  • Dial Size (in)3
  • Dial / Case Material304 Stainless Steel
  • Stem Length (in)2.5
  • Stem Diameter (in)0.25
  • Stem Material304 Stainless Steel
  • Connection0.5\" NPT(M)
  • DescriptionBimetal Thermometer 3\" 200/1000F 2.5\" L, 1/2\" NPT Adjustable Angle
More About this Item
Our 3" dial and 5" dial process grade bimetal thermometers offer the convenience of an allangle swivel connection. Additionally, the hermetically- sealed case offers protection from weather and dust, and is guaranteed against fogging up. An inert gel is used to minimize
pointer oscillation. These thermometers are guaranteed for seven years.
WIKA Bimetal thermometers are manufactured to ASME B40.3 standards.
Additional ranges and stem lengths are available.