Wide-Range Relief Valve, Polypropylene, 1/2" NPT(F)
Representative image only.
Item # WZ-01350-66
Prevent leaks with the emergency shutdown design
  • Three-seal system vents liquid to warn of approaching maintenance
PRODUCT OPTIONS
£336.96GBP / Each
£404.35GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/2" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/2" NPT(F)
  • Max Pressure (PSI)210
  • Max Pressure (bar)14.5
  • Wetted MaterialsPolypropylene and Viton
  • Media CompatibilityLiquids
  • DescriptionWide-Range Relief Valve, Polypropylene, 1/2" NPT(F)
Key Features
  • Three-seal system vents liquid to warn of approaching maintenance
More About this Item
Valves with connections of 1/4" or 1/2" have a diaphragm made of Viton® and withstand a maximum inlet pressure of 210 psig. All other valves have a PTFE shaft design and withstand a maximum inlet pressure of 140 psig. The shaft design prevents emergency shutdown with a three-seal system that will vent liquid to warn of approaching maintenance. All valves are recommended for liquid applications.