DWK Life Sciences (Wheaton) W225301 Vial Crimping Pliers/Crimper for 11 mm Seals

Representative image only.
DWK Life Sciences (Wheaton) Mfr # W225301 – Item # WZ-98800-67
Ergonomic design reduces hand fatigue
  • Suitable for use with 11 mm seals
  • 2 year warranty

£554.94GBP / Each
£665.93GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Cap Diameter (mm)11
  • DescriptionVial Crimping Pliers/Crimper for 11 mm Seals
  • Warranty2 year
Two year warranty

Key Features

  • Suitable for use with 11 mm seals
  • 2 year warranty

More About this Item

Manufactured from sturdy, cast aluminum and is lightweight and easy to use. Polished, precision sealing jaws are tapered to allow crimping vials in racks or trays. An adjustable stop enables you to achieve a tight, uniform seal every time with a variety of vials.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat