DWK Life Sciences (Wheaton) Soxhlet extraction glassware systems, 250 mL
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 416005 – Item # WZ-99036-20
Extract soluble components from a solid sample into an organic solvent
  • Soxhlets are made of low extractable borosilicate glass and come complete with extractor, condenser, and flask
  • Model 99036-00 has screw thread connections
  • Models 99036-10 and -20 have ground glass joints
  • An extraction thimble is required to support the sample within the extractor
  • 1 year warranty
PRODUCT OPTIONS
£759.78GBP / Each
£911.74GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)250
  • Condenser TypeAllihn
  • Flask TypeFlat-bottom
  • Flask Joint Size24/40
  • Condenser Joint Size24/40
  • DescriptionSoxhlet extraction glassware systems, 250 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • Soxhlets are made of low extractable borosilicate glass and come complete with extractor, condenser, and flask
  • Model 99036-00 has screw thread connections
  • Models 99036-10 and -20 have ground glass joints
  • An extraction thimble is required to support the sample within the extractor
  • 1 year warranty
More About this Item
Place a soil sample in the thimble and heat the solvent contained in the flask. As the solvent vapors rise (passing through the extraction thimble), they enter the water-cooled condenser, and reliquify. When the liquid level in the extractor reaches the top of the siphon tube, siphoning action returns the extract-enriched solvent to the flask.