DWK Life Sciences (Wheaton) 357844 Disposable Micro-Tissue Grinder, Glass, 0.1 mL; 2/Cs
Representative image only.
DWK Life Sciences (Wheaton) Mfr # 357844 – Item # WZ-04468-38
Designed for extremely precise work and delicate hand operation
  • Made from low extractable borosilicate glass
  • 1 year warranty
£213.09GBP / Case of 2
£255.71GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)0.1
  • Capacity (Liters)0.0001
  • MaterialGlass
  • Grinding Chamber (mmDia)4
  • Grinding Chamber (mmL)7.5
  • Overall Length (mm)100
  • DescriptionDisposable Micro-Tissue Grinder, Glass, 0.1 mL; 2/Cs
  • Warranty1 year
1 Year Warranty
Key Features
  • Made from low extractable borosilicate glass
  • 1 year warranty
More About this Item
Similar to the Potter-Elvehjem tissue grinders, but for micro samples. No washing is necessary; just dispose of the tissue grinder after use. Ideal for radioisotopes. The small grinding area yields a high percentage of sample recovery. Grinding chamber clearance is 0.004" to 0.006".