VistaLab Reagent Reservoir Cooler with Lid, for 100 mL Reservoir, Sterile

Representative image only.
VistaLab Mfr # 3054-5000 – Item # WZ-06198-82
Chill or freeze quickly and maintain reagents at <5°C for up to 1 hour
  • Reusable reagent reservoir cooler is compatible with VistaLab 100 mL reagent reservoirs
  • Manufactured with non-toxic gel encased in heavy duty PVC
  • 1 year warranty

£44.72GBP / Each
£53.66GBP (incl VAT)
Often Bought With…
VistaLab™ Reagent Reservoirs, Polystyrene
£77.84 - £102.90GBP / Case of 100
£93.41 - £123.48GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • SterileYes
  • Wetted MatterialGel Encased in Heavy Duty PVC
  • Compatible withVistaLab 100 mL Reagent Reservoirs
  • DescriptionReagent Reservoir Cooler with Lid, for 100 mL Reservoir, Sterile
1 Year Warranty

Key Features

  • Reusable reagent reservoir cooler is compatible with VistaLab 100 mL reagent reservoirs
  • Manufactured with non-toxic gel encased in heavy duty PVC
  • 1 year warranty

More About this Item

The reusable, nontoxic gel cooler preserves the integrity of reagents and extends working time with full reservoir cooling. Custom fit for the 100-mL reagent reservoirs, the cooler will maintain your reagent at <41°F (5°C) for up to two hours. Order reagent reservoirs separately.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat