VistaLab VistaTip™ Pipette Tips, Sterile, Individually Wrapped, RNase/DNase Free, 250 µL; 200/Pk
Representative image only.
VistaLab Mfr # 2027 – Item # WZ-06199-75
Maximize performance and benefits of your pipette
  • Tips made of 100% pure virgin polypropylene
PRODUCT OPTIONS
£80.07GBP / Pkg of 200
£96.08GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (µL)250
  • Min Volume (µL)2
  • Overall Length (in)2.16
  • Overall Length (mm)54.8
  • Tip StyleSmall; RNase/DNase Free
  • Wetted MatterialPolypropylene
  • Package OfIndividually Wrapped; 200/Pk
  • Compatible withOvation ESC, E8, E12, QS, M, F1, F2 & All MLA model Pipettors
  • DescriptionPipette Tips, Sterile, Individually Wrapped, RNase/DNase Free, 250 µL; 200/Pk
Key Features
  • Tips made of 100% pure virgin polypropylene
More About this Item
VistaTip™ polypropylene (PP) pipette tips are pre-sterilized and individually wrapped to reduce cross-contamination. Choose from standard or certified RNase/DNase free non-pyrogenic tips. Tips are compatible with VistaLab ™ Ovation and MLA model pipettors.
Tips are certified RNase/DNase free and non-pyrogenic.