VistaLab 1103 Fixed-Volume Pipette, Silver, 30 uL; Each
Representative image only.
VistaLab Mfr # 1103 – Item # WZ-06198-01
Ideal for a broad range of laboratory procedures requiring precision and accuracy
  • Choose from fixed volumes between 5 µL up to 1000 µL
  • Simple single-stroke procedure
  • Lower spring force minimizes muscle aches due to repetitive motion
  • Beaker features a 4000 mL capacity
  • Designed with graduation range from 500 to 3500 mL in 500 mL intervals
  • Limited Lifetime Warranty
PRODUCT OPTIONS
£207.97GBP / Each
£249.56GBP (incl VAT)
£90.44 - £142.45GBP / Each
£108.53 - £170.94GBP (incl VAT)
for an additional
£90.44GBP / Each
£108.53GBP (incl VAT)
for an additional
£142.45GBP / Each
£170.94GBP (incl VAT)
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought With…
VistaLab™ VistaRak™ Pipette Tips
£67.04 - £169.15GBP / Case of 960
£80.45 - £202.98GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (µL)30
  • Max Volume (mL)0.03
  • Max Dispensing (mL)0.03
  • Accuracy (%)±3.00%
  • Precision (%)±0.93%
  • Overall Length (in)7.48
  • Overall Length (mm)190
  • Color CodeSilver
  • Compatible withVistaLab \"Small\" tips
  • DescriptionFixed-Volume Pipette, Silver, 30 uL; Each
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Choose from fixed volumes between 5 µL up to 1000 µL
  • Simple single-stroke procedure
  • Lower spring force minimizes muscle aches due to repetitive motion
  • Beaker features a 4000 mL capacity
  • Designed with graduation range from 500 to 3500 mL in 500 mL intervals
  • Limited Lifetime Warranty
More About this Item
The MLA Precision pipettes are calibrated “to deliver” (TD) pipettes, use air-displacement, and require manual de-tipping. They come factory calibrated and tested at 21.5°C ±2°C using distilled water. Pipettors consistently deliver the calibrated volume whenever the plunger is fully depressed. Calibration volume can be changed using the calibration key (included).
Pipettors are color-coded for easy identification and come with the stated volume engraved onto the piston assembly. Engineered with aluminum alloy and stainless steel, these pipettors withstand years of use and provide exceptional performance for a wide range of fluids, including dense or viscous samples.