VESPel Graphite Short Ferrules Compatible with Agilent Inlets, 60% Vespel and 40% Graphite, 1/16" to 0.5 mm; 10/Pk
Representative image only.
VESPel Mfr # 213165 – Item # WZ-01998-68
Ideal for GC/MS interface applications
  • Manufactured of 60% Vespel and 40% Graphite
  • 1 month warranty
PRODUCT OPTIONS
£68.21GBP / Pkg of 10
£81.85GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGraphite Short Ferrules Compatible with Agilent Inlets, 1/16\" to 0.5 mm
  • Warranty1 month
1 Month Warranty
Key Features
  • Manufactured of 60% Vespel and 40% Graphite
  • 1 month warranty
More About this Item
Vespel®/Graphite Ferrules are one-piece design requiring no back ferrule. Maximum temperature: 400°C. Recommended for GC/MS interface applications. Reusable – does not seize into column.