VESPel Graphite Straight Ferrules for Packed Columns, 60% Vespel and 40% Graphite, 1/16"; 10/Pk
Representative image only.
VESPel Mfr # 213100 – Item # WZ-01993-50
Ideal for GC/MS interface applications
  • Manufactured of 60% Vespel and 40% Graphite
PRODUCT OPTIONS
£63.96GBP / Pkg of 10
£76.75GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Description/Graphite Straight Ferrules for Packed Columns, 1/16\"
Key Features
  • Manufactured of 60% Vespel and 40% Graphite
More About this Item
Vespel®/Graphite Ferrules are one-piece design requiring no back ferrule. Maximum temperature: 400°C. Recommended for GC/MS interface applications. Reusable – does not seize into column.