This item is now discontinued. Please browse similar items
Velp KjTabs Catalyst Tablets, VKPC; 1000/BX
Representative image only.
Velp Mfr # A00000275 – Item # WZ-86580-06
Speed up digestions
  • Compatible with any digestion system Wide variety of catalyst formulae work with most samples
  • Formulation: 4.5 g K2SO4 + 0.5 g CuSO4 x 5 H2O
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Materials of Construction4.5 g K2SO4 + 0.5 g CuSO4 x 5 H2O
  • DescriptionCatalyst Tablets, VKPC; 1000/BX
Two year warranty
Key Features
  • Compatible with any digestion system Wide variety of catalyst formulae work with most samples
  • Formulation: 4.5 g K2SO4 + 0.5 g CuSO4 x 5 H2O
  • 2 year warranty
More About this Item
Use with the Velp DK and DKL Digestion Systems, or any other similar system.